Isoparaffin G analytisches Reagens

Lieferant: PanReac AppliChem
Gefahr


Filtered through a 0.2 µm filter.

125273.1612 125273.1214
APPC125273.1612EA 223 CHF
APPC125273.1612 APPC125273.1214
Isoparaffin G analytisches Reagens
Isoparaffin
CAS-Nummer: 90622-57-4
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR