tetra-Natriumdiphosphat Decahydrat ≥99,0% (durch Acidimetrie) zur Analyse

Lieferant: PanReac AppliChem

Synonyms: Sodium pyrophosphate decahydrate, tetra-Sodium pyrophosphate decahydrate, Sodium pyrophosphate tetrabasic decahydrate, TSPP

121710.1211
APPC121710.1211EA 211 CHF
APPC121710.1211
tetra-Natriumdiphosphat Decahydrat ≥99,0% (durch Acidimetrie) zur Analyse
tetra-Natriumdiphosphat Decahydrat
Formel: Na₄P₂O₇·10H₂O
Melting Pt: 79 °C
Dichte: 1.82
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149200
CAS-Nummer: 13472-36-1
EINECS: 231-767-1

Order Now

Spezifikation Testergebnisse

Minimum Assay (Acidimetry) 99.0%
pH of 5% Solution 9.5 - 10.7
Insoluble Matter in H₂O ≤0.01%
Chloride (Cl) ≤0.005%
Sulphate (SO₄) ≤0.005%
Carbonate Passes Test
Nitrate (NO₃) ≤0.003%
As ≤0.0003%
Ca ≤001%
Cu ≤0.0005%
Fe ≤0.0005%
MG ≤0.01%
Ni ≤0.0005%
PB ≤0.0005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR