Methyl 4-(3-fluorophenyl)butanoate 95%

Lieferant: Apollo Scientific

PC907969-1G
APOSPC907969-1GEA 453 CHF
APOSPC907969-1G
Methyl 4-(3-fluorophenyl)butanoate 95%
Methyl 4-(3-fluorophenyl)butanoate
Formel: C₁₁H₁₃FO₂
MW: 196,218 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD14635874
CAS-Nummer: 134464-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR