1-(2,2,2-Trifluorethyl)pyrrolidin 95%

Lieferant: Apollo Scientific

PC906398-1G
APOSPC906398-1GEA 584 CHF
APOSPC906398-1G
1-(2,2,2-Trifluorethyl)pyrrolidin 95%
1-(2,2,2-Trifluoroethyl)pyrrolidine
Formel: C₆H₁₀F₃N
MW: 153.15 g/mol
Lager Temperatur: Raumtemperatur
CAS-Nummer: 1378304-83-6

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR