4-Chloro-7-fluoro-2-methylquinazoline 97%

Lieferant: Apollo Scientific

PC905284-5G
APOSPC905284-5GEA 236 CHF
APOSPC905284-5G
4-Chloro-7-fluoro-2-methylquinazoline 97%
4-Chloro-7-fluoro-2-methylquinazoline
Formel: C₉H₆ClFN₂
MW: 196,609 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD12024899
CAS-Nummer: 1206694-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR