N-Acetyl 5-fluoro-2-methylaniline 98%

Lieferant: Apollo Scientific

Synonyms: 2-Acetamido-4-fluorotoluene

PC902344-25G
APOSPC902344-25GEA 168 CHF
APOSPC902344-25G
N-Acetyl 5-fluoro-2-methylaniline 98%
N-Acetyl 5-fluoro-2-methylaniline
Formel: C₉H₁₀FNO
MW: 167.18 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00045064
CAS-Nummer: 366-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR