4-Bromo-2-fluoro-5-(trifluoromethyl)phenylboronic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyms: 4-Bromo-2-fluoro-5(trifluoromethyl)phenylboronic acid, 4-Bromo-2-fluoro-5-(trifluoromethyl)benzeneboronic acid

PC900987-250MG PC900987-1G PC900987-5G
APOSPC900987-250MGEA 101 CHF
APOSPC900987-250MG APOSPC900987-1G APOSPC900987-5G
4-Bromo-2-fluoro-5-(trifluoromethyl)phenylboronic acid 95%
4-Bromo-2-fluoro-5-(trifluoromethyl)phenylboronic acid
Formel: C₇H₄BBrF₄O₂
MW: 286.81 g/mol
MDL Number: MFCD20441807
CAS-Nummer: 1451393-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR