2-Brom-5-fluorphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyms: (2-Bromo-5-fluorophenyl)boronic acid

PC8649-1G
APOSPC8649-1GEA 141 CHF
APOSPC8649-1G
2-Brom-5-fluorphenylboronsäure
2-Brom-5-fluorphenylboronsäure
Formel: C₆H₅BBrFO₂
MW: 218,82 g/mol
Lager Temperatur: Kühlschrank
MDL Number: MFCD08690232
CAS-Nummer: 959996-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR