1,3-Dichlor-7-fluorisochinolin 98%

Lieferant: Apollo Scientific

PC7422-1G
APOSPC7422-1GEA 367 CHF
APOSPC7422-1G
1,3-Dichlor-7-fluorisochinolin 98%
1,3-Dichloro-7-fluoroisoquinoline
Formel: C₉H₄Cl₂FN
MW: 216,0392 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD09763685
CAS-Nummer: 941294-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR