4,4-Difluorocyclohexylamine hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyms: 4,4-Difluorocyclohexanamine hydrochloride

PC7317-5G PC7317-1G PC7317-250MG PC7317-25G
APOSPC7317-5GEA 161 CHF
APOSPC7317-5G APOSPC7317-1G APOSPC7317-250MG APOSPC7317-25G
4,4-Difluorocyclohexylamine hydrochloride 98%
4,4-Difluorocyclohexylamine hydrochloride
Formel: C₆H₁₂ClF₂N
MW: 171.62 g/mol
Melting Pt: 106…107 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD06246881
CAS-Nummer: 675112-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR