2-Fluoro-6-methylphenol 97%

Lieferant: Apollo Scientific
Achtung

Synonyms: 3-Fluoro-2-hydroxytoluene

PC5866-500MG
APOSPC5866-500MGEA 124 CHF
APOSPC5866-500MG
2-Fluoro-6-methylphenol 97%
2-Fluoro-6-methylphenol
Formel: C₇H₇FO
MW: 126.128 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD12026463
CAS-Nummer: 443-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR