(1-Bromo)perfluoro-2,5,8-trioxanonane 98%

Lieferant: Apollo Scientific
Achtung

PC5539-100G PC5539-25G PC5539-5G PC5539-1G
APOSPC5539-100GEA 0 CHF
APOSPC5539-100G APOSPC5539-25G APOSPC5539-5G APOSPC5539-1G
(1-Bromo)perfluoro-2,5,8-trioxanonane 98%
(1-Bromo)perfluoro-2,5,8-trioxanonane
Formel: C₆BrF₁₃O₃
MW: 446.95 g/mol
Siedepunkt: 100 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD03094295
CAS-Nummer: 330562-45-3

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR