8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one

Lieferant: Apollo Scientific
Achtung

PC53585-250MG
APOSPC53585-250MGEA 131 CHF
APOSPC53585-250MG
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one
Formel: C₇H₃ClF₃N₃O
MW: 237,566 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD12406875
CAS-Nummer: 1428532-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR