3-(Difluoromethyl)-5-methyl-1H-indole-2-carboxylic acid

Lieferant: Apollo Scientific

PC520075-1G
APOSPC520075-1GEA 3290 CHF
APOSPC520075-1G
3-(Difluoromethyl)-5-methyl-1H-indole-2-carboxylic acid
3-(Difluoromethyl)-5-methyl-1H-indole-2-carboxylic acid
Formel: C₁₁H₉F₂NO₂
MW: 225.191 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD29767559
CAS-Nummer: 2092578-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR