3-Fluoro-2-nitrophenacyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC501205-1G PC501205-250MG PC501205-5G
APOSPC501205-1GEA 97.5 CHF
APOSPC501205-1G APOSPC501205-250MG APOSPC501205-5G
3-Fluoro-2-nitrophenacyl bromide 95%
3-Fluoro-2-nitrophenacyl bromide
Formel: C₈H₅BrFNO₃
Lager Temperatur: Raumtemperatur
MDL Number: MFCD23711759
CAS-Nummer: 1427431-58-0
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR