2-Fluoro-5-nitrobenzenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

PC500491-1G PC500491-250MG PC500491-5G
APOSPC500491-1GEA 240 CHF
APOSPC500491-1G APOSPC500491-250MG APOSPC500491-5G
2-Fluoro-5-nitrobenzenesulphonyl chloride
2-Fluoro-5-nitrobenzenesulphonyl chloride
Formel: C₆H₃ClFNO₄S
Lager Temperatur: Raumtemperatur
MDL Number: MFCD08443561
CAS-Nummer: 713-21-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR