α-Methyl-3-(trifluoromethyl)benzylamine 98%

Lieferant: Apollo Scientific
Achtung

Synonyms: 1-(3-(Trifluoromethyl)phenyl)ethanamine, 1-[3-(Trifluoromethyl)phenyl]ethylamine, 3-(1-Aminoethyl)benzotrifluoride

PC4459-1G PC4459-250MG
APOSPC4459-1GEA 59.2 CHF
APOSPC4459-1G APOSPC4459-250MG
α-Methyl-3-(trifluoromethyl)benzylamine 98%
α-Methyl-3-(trifluoromethyl)benzylamine
Formel: C₉H₁₀F₃N
MW: 189.178 g/mol
Siedepunkt: 69.4…70.8 °C (45 mm Hg)
Flash-Pt: 72 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD04038877
CAS-Nummer: 59382-36-4

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR