2-Nitro-3-(2-(trifluoromethyl)-phenyl)naphthalene

Lieferant: Apollo Scientific

PC440013-100MG
APOSPC440013-100MGEA 2150 CHF
APOSPC440013-100MG
2-Nitro-3-(2-(trifluoromethyl)-phenyl)naphthalene
2-Nitro-3-(2-(trifluoromethyl)-phenyl)naphthalene
Formel: C₁₇H₁₀F₃NO₂
MW: 317,262 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD28154812
CAS-Nummer: 1450995-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR