2-Fluor-5-(trifluormethyl)benzylamin 97%

Lieferant: Apollo Scientific
Gefahr

PC4374V-1G
APOSPC4374V-1GEA 27.9 CHF
APOSPC4374V-1G
2-Fluor-5-(trifluormethyl)benzylamin 97%
2-Fluor-5-(trifluormethyl)benzylamin
Formel: C₈H₇F₄N
MW: 193.14 g/mol
Siedepunkt: 185
Melting Pt: 46
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00061162
CAS-Nummer: 199296-61-2
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR