3-(Trifluoroacetamido)pyrrolidine hydrochloride

Lieferant: Apollo Scientific

PC430507-1G
APOSPC430507-1GEA 172 CHF
APOSPC430507-1G
3-(Trifluoroacetamido)pyrrolidine hydrochloride
3-(Trifluoroacetamido)pyrrolidine hydrochloride
Formel: C₆H₉F₃N₂O
MW: 218.60 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01090944
CAS-Nummer: 84424-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR