3-Acetamido-2-nitrophenyl 4-(dimethylamino)-3,5-difluorobenzoate

Lieferant: Apollo Scientific

PC430220-1G
APOSPC430220-1GEA 786 CHF
APOSPC430220-1G
3-Acetamido-2-nitrophenyl 4-(dimethylamino)-3,5-difluorobenzoate
3-Acetamido-2-nitrophenyl 4-(dimethylamino)-3,5-difluorobenzoate
Formel: C₁₇H₁₅F₂N₃O₅
MW: 379,315 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD27978603
CAS-Nummer: 1864063-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR