2-(3,4-Difluorophenyl)-4'-fluoroacetophenone

Lieferant: Apollo Scientific

PC421006-1G
APOSPC421006-1GEA 436 CHF
APOSPC421006-1G
2-(3,4-Difluorophenyl)-4'-fluoroacetophenone
2-(3,4-Difluorophenyl)-4'-fluoroacetophenone
Formel: C₁₄H₉F₃O
MW: 250,216 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD24968600
CAS-Nummer: 1507249-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR