5-{[4-Bromo-3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid

Lieferant: Apollo Scientific

PC410451-1G
APOSPC410451-1GEA 279 CHF
APOSPC410451-1G
5-{[4-Bromo-3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid
5-{[4-Bromo-3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid
Formel: C₁₁H₇BrF₄N₂O₃
MW: 371.082 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD03419729
CAS-Nummer: 1006447-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR