1-(Difluoromethyl)-3-methyl-1H-pyrazole-5-carboxylic acid

Lieferant: Apollo Scientific

PC410370-1G
APOSPC410370-1GEA 1300 CHF
APOSPC410370-1G
1-(Difluoromethyl)-3-methyl-1H-pyrazole-5-carboxylic acid
1-(Difluoromethyl)-3-methyl-1H-pyrazole-5-carboxylic acid
Formel: C₆H₆F₂N₂O₂
MW: 176.12 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD04970129
CAS-Nummer: 1006319-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR