1-[(2-Fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid

Lieferant: Apollo Scientific

PC410357-1G
APOSPC410357-1GEA 776 CHF
APOSPC410357-1G
1-[(2-Fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
1-[(2-Fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
Formel: C₁₁H₉FN₂O₃
MW: 236,199 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD04050603
CAS-Nummer: 1005662-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR