5-(Pentafluoroethyl)pyridin-2-amine

Lieferant: Apollo Scientific

PC408845-1G
APOSPC408845-1GEA 2000 CHF
APOSPC408845-1G
5-(Pentafluoroethyl)pyridin-2-amine
5-(Pentafluoroethyl)pyridin-2-amine
Formel: C₇H₅F₅N₂
MW: 212,12 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD28122893
CAS-Nummer: 1427789-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR