5-Fluoro-2-(piperidin-4-ylmethoxy)-pyrimidine hydrochloride

Lieferant: Apollo Scientific

PC408363-500MG
APOSPC408363-500MGEA 663 CHF
APOSPC408363-500MG
5-Fluoro-2-(piperidin-4-ylmethoxy)-pyrimidine hydrochloride
5-Fluoro-2-(piperidin-4-ylmethoxy)-pyrimidine hydrochloride
Formel: C₁₀H₁₄FN₃O·ClH
MW: 247.697 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD21098930
CAS-Nummer: 1353965-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR