(5-Bromo-2-(difluoromethoxy)pyridin-3-yl)methanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC400162-100MG PC400162-250MG
APOSPC400162-100MGEA 237 CHF
APOSPC400162-100MG APOSPC400162-250MG
(5-Bromo-2-(difluoromethoxy)pyridin-3-yl)methanamine hydrochloride
(5-Bromo-2-(difluoromethoxy)pyridin-3-yl)methanamine hydrochloride
Formel: C₇H₇BrF₂N₂O·ClH
MW: 289.505 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD31621144
CAS-Nummer: 2231675-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR