4'-Fluoracetophenon 99%

Lieferant: Apollo Scientific
Achtung

Synonyms: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone

PC3380-100G PC3380-1KG PC3380-250G
APOSPC3380-100GEA 52.2 CHF
APOSPC3380-100G APOSPC3380-1KG APOSPC3380-250G
4'-Fluoracetophenon 99%
4'-Fluoracetophenon
Formel: C₈H₇FO
MW: 138.14 g/mol
Siedepunkt: 196 °C
Dichte: 1.138 g/cm³ (25 °C)
Flash-Pt: 71 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000354
CAS-Nummer: 403-42-9
EINECS: 206-960-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR