PTFE

Lieferant: Apollo Scientific
Achtung

Synonyms: Polytetrafluoroethylene, Teflon

PC3329-5KG PC3329-5G
APOSPC3329-5KGEA 0 CHF
APOSPC3329-5KG APOSPC3329-5G
PTFE
PTFE

60 wt % dispersion in water; 0.05-0.5 micron particles.

Formel: (CF₂CF₂)n
Dichte: 1.5 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00084455
CAS-Nummer: 9002-84-0

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR