4-Fluoro-5-nitrobenzene-1,2-diamine

Lieferant: Apollo Scientific
Achtung

Synonyms: 1,2-Diamino-3-fluoro-4-nitrobenzene, 4,5-Diamino-2-fluoronitrobenzene

PC31311-1G
APOSPC31311-1GEA 138 CHF
APOSPC31311-1G
4-Fluoro-5-nitrobenzene-1,2-diamine
4-Fluoro-5-nitrobenzene-1,2-diamine
Formel: C₆H₆FN₃O₂
MW: 171,129 g/mol
Melting Pt: 195…196 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00828924
CAS-Nummer: 113269-06-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR