2'-Fluoro-3'-methyl-5'-(trifluoromethyl)acetophenone 97%

Lieferant: Apollo Scientific

PC303384-1G PC303384-250MG
APOSPC303384-1GEA 780 CHF
APOSPC303384-1G APOSPC303384-250MG
2'-Fluoro-3'-methyl-5'-(trifluoromethyl)acetophenone 97%
2'-Fluoro-3'-methyl-5'-(trifluoromethyl)acetophenone
Formel: C₁₀H₈F₄O
MW: 220.164 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD31645673
CAS-Nummer: 2055275-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR