4-Chlor-2-(trifluormethyl)benzylamin 97%

Lieferant: Apollo Scientific

PC303078-5G
APOSPC303078-5GEA 94 CHF
APOSPC303078-5G
4-Chlor-2-(trifluormethyl)benzylamin 97%
4-Chlor-2-(trifluormethyl)benzylamin
Formel: C₈H₇ClF₃N
MW: 209.6 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD06213291
CAS-Nummer: 771583-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR