3-Methyl-5-(trifluoromethoxy)cinnamic acid 98%

Lieferant: Apollo Scientific

PC302310-1G
APOSPC302310-1GEA 177 CHF
APOSPC302310-1G
3-Methyl-5-(trifluoromethoxy)cinnamic acid 98%
3-Methyl-5-(trifluoromethoxy)cinnamic acid
Formel: C₁₁H₉F₃O₃
MW: 246,183 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD09025375
CAS-Nummer: 1005378-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR