(4'-Fluoro-1,1'-biphenyl-4-yl)methanol

Lieferant: Apollo Scientific

PC300617-1G
APOSPC300617-1GEA 297 CHF
APOSPC300617-1G
(4'-Fluoro-1,1'-biphenyl-4-yl)methanol
(4'-Fluoro-1,1'-biphenyl-4-yl)methanol
Formel: C₁₃H₁₁FO
MW: 202,224 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01862513
CAS-Nummer: 147497-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR