1-Fluorocyclohexanecarboxylic acid

Lieferant: Apollo Scientific
Achtung

PC2955-250MG
APOSPC2955-250MGEA 268 CHF
APOSPC2955-250MG
1-Fluorocyclohexanecarboxylic acid
1-Fluorocyclohexanecarboxylic acid
Formel: C₇H₁₁FO₂
MW: 146.16 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD08751334
CAS-Nummer: 117169-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR