1-Methyl-5-(morpholin-4-yl)-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile, tech

Lieferant: Apollo Scientific

PC28110-1G
APOSPC28110-1GEA 1880 CHF
APOSPC28110-1G
1-Methyl-5-(morpholin-4-yl)-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile, tech
1-Methyl-5-(morpholin-4-yl)-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile
Formel: C₁₀H₁₁F₃N₄O
MW: 260,216 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00141044
CAS-Nummer: 318517-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR