3-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-4-yl]-3-oxopropanenitrile, tech

Lieferant: Apollo Scientific

PC28055-1G
APOSPC28055-1GEA 1880 CHF
APOSPC28055-1G
3-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-4-yl]-3-oxopropanenitrile, tech
3-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-4-yl]-3-oxopropanenitrile
Formel: C₈H₆F₃N₃O
MW: 217,148 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD03012277
CAS-Nummer: 956779-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR