1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-4,5-dimethyl-2,3-dihydro-1H-pyrazol-3-one, tech

Lieferant: Apollo Scientific

PC28024-1G
APOSPC28024-1GEA 1880 CHF
APOSPC28024-1G
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-4,5-dimethyl-2,3-dihydro-1H-pyrazol-3-one, tech
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-4,5-dimethyl-2,3-dihydro-1H-pyrazol-3-one
Formel: C₁₁H₉ClF₃N₃O
MW: 291,657 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00172554
CAS-Nummer: 338769-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR