2-Oxo-6-[3-(trifluoromethyl)phenyl]-1,2-dihydropyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

PC27950-1G
APOSPC27950-1GEA 1050 CHF
APOSPC27950-1G
2-Oxo-6-[3-(trifluoromethyl)phenyl]-1,2-dihydropyridine-3-carbonitrile, tech
2-Oxo-6-[3-(trifluoromethyl)phenyl]-1,2-dihydropyridine-3-carbonitrile
Formel: C₁₃H₇F₃N₂O
MW: 264,203 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00215071
CAS-Nummer: 140692-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR