4-Brom-2-fluorbenzonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyms: 1-Brom-4-cyan-3-fluorbenzene

PC2665-100G
APOSPC2665-100GEA 151 CHF
APOSPC2665-100G
4-Brom-2-fluorbenzonitril 98%
4-Brom-2-fluorbenzonitril
Formel: C₇H₃BrFN
MW: 200.01 g/mol
Siedepunkt: 238…239 °C
Melting Pt: 69…72 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00143264
CAS-Nummer: 105942-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR