Methyl-4-amino-2,6-difluorobenzoate 98%

Lieferant: Apollo Scientific

Synonyms: Methyl 4-amino-2,6-difluorobenzoate

PC201187-100MG PC201187-1G PC201187-250MG
APOSPC201187-100MGEA 121 CHF
APOSPC201187-100MG APOSPC201187-1G APOSPC201187-250MG
Methyl-4-amino-2,6-difluorobenzoate 98%
Methyl-4-amino-2,6-difluorobenzoate
Formel: C₈H₇F₂NO₂
MW: 187.14 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD16038807
CAS-Nummer: 191478-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR