2,4-Dichloro-5-fluoropyridine 98%

Lieferant: Apollo Scientific

PC201185-100MG PC201185-1G PC201185-250MG
APOSPC201185-100MGEA 142 CHF
APOSPC201185-100MG APOSPC201185-1G APOSPC201185-250MG
2,4-Dichloro-5-fluoropyridine 98%
2,4-Dichloro-5-fluoropyridine
Formel: C₅H₂Cl₂FN
MW: 165.98 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD18257011
CAS-Nummer: 189281-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR