1-Brom-4-fluor-3-nitrobenzol 98%

Lieferant: Apollo Scientific
Achtung

Synonyms: 4-Bromo-1-fluoro-2-nitrobenzene, 5-Bromo-2-fluoronitrobenzene

PC1428D-100G PC1428D-25G PC1428D-500G
APOSPC1428D-100GEA 31.4 CHF
APOSPC1428D-100G APOSPC1428D-25G APOSPC1428D-500G
1-Brom-4-fluor-3-nitrobenzol 98%
1-Bromo-4-fluoro-3-nitrobenzene
Formel: C₆H₃BrFNO₂
MW: 220 g/mol
Siedepunkt: 240…241 °C
Melting Pt: 18…19 °C
Dichte: 1.786 g/cm³ (25 °C)
Flash-Pt: >110 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00129165
CAS-Nummer: 364-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR