2-Brom-4-(trifluormethyl)benzoesäure 97+%

Lieferant: Apollo Scientific
Achtung

Synonyms: 2-Bromo-4-(trifluoromethyl)benzoic acid, 2-Bromo-4-(trifluoromethl)benzoic acid

PC11107-5G
APOSPC11107-5GEA 76.6 CHF
APOSPC11107-5G
2-Brom-4-(trifluormethyl)benzoesäure 97+%
2-Brom-4-(trifluormethyl)benzoesäure
Formel: C₈H₄BrF₃O₂
MW: 269.02 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD07368837
CAS-Nummer: 328-89-2
EINECS: 206-339-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR