Diethyl 2-(4-fluorophenyl)malonate

Lieferant: Apollo Scientific
Achtung

Synonyms: Diethyl 2-(4-fluorophenyl)propane-1,3-dioate

PC10099-1G
APOSPC10099-1GEA 55.7 CHF
APOSPC10099-1G
Diethyl 2-(4-fluorophenyl)malonate
Diethyl 2-(4-fluorophenyl)malonate
Formel: C₁₃H₁₅FO₄
Siedepunkt: 161…162 °C (15 mm Hg)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD04039208
CAS-Nummer: 2965-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR