1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether 97%

Lieferant: Apollo Scientific
Achtung

Synonyms: 2,2,3,3,4,4,5,5-Octafluoropentyl 1,1,2,2-Tetrafluoroethyl Ether, 1,1,2,2,3,3,4,4-Octafluoro-5-(1,1,2,2-tetrafluoroethoxy)pentane

PC10021-25G PC10021-100G PC10021-500G
APOSPC10021-25GEA 95.8 CHF
APOSPC10021-25G APOSPC10021-100G APOSPC10021-500G
1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether 97%
1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether
Formel: C₇H₄F₁₂O
MW: 332.09 g/mol
Siedepunkt: 133 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00155904
CAS-Nummer: 16627-71-7

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR