4-Brom-3-fluorthiophenol 95%

Lieferant: Apollo Scientific
Achtung

PC01517-1G PC01517-5G PC01517-250MG
APOSPC01517-1GEA 332 CHF
APOSPC01517-1G APOSPC01517-5G APOSPC01517-250MG
4-Brom-3-fluorthiophenol 95%
4-Brom-3-fluorthiophenol
Formel: C₆H₄BrFS
MW: 207.06 g/mol
Lager Temperatur: Kühlschrank
MDL Number: MFCD09475476
CAS-Nummer: 942473-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR