Methyl-2-methylimidazo[1,2-a]pyridin-3-carboxylat 95%

Lieferant: Apollo Scientific

Synonyms: Methyl 2-methylimidazo[1,2-a]pyridine-3-carboxylate

OR965437-1G
APOSOR965437-1GEA 117 CHF
APOSOR965437-1G
Methyl-2-methylimidazo[1,2-a]pyridin-3-carboxylat 95%
Methyl-2-methylimidazo[1,2-a]pyridin-3-carboxylat
Formel: C₁₀H₁₀N₂O₂
MW: 190.2000 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD15203666
CAS-Nummer: 943112-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR