2-(6-Chloropyrazin-2-ylamino)acetic acid 95%

Lieferant: Apollo Scientific

OR963015-1G OR963015-250MG
APOSOR963015-1GEA 688 CHF
APOSOR963015-1G APOSOR963015-250MG
2-(6-Chloropyrazin-2-ylamino)acetic acid 95%
2-(6-Chloropyrazin-2-ylamino)acetic acid
Formel: C₆H₆ClN₃O₂
Lager Temperatur: Raumtemperatur
MDL Number: MFCD12068353
CAS-Nummer: 1159822-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR